missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Qorpak™ Graduated Glass Bottle Beakers with Black Phenolic Pulp/Vinyl Caps

For mixing, measuring and storage

Manufacturer:  Qorpak™ GLC01521

 View more versions of this product

Catalog No. 02992077



Description

Description

  • Designed with slight shoulder for easy pouring and handling
  • Molded graduations in mL and ounces
  • Made of clear glass
  • For use with biological and pathological specimens
  • For storage of industrial laboratory chemicals and reagents

Specifications

Specifications

4 oz., 120mL
4 oz.
Pulp/Vinyl
2.01 in.
Glass
144/Cs.
48-400
102mm
120mL
Phenolic
Clear
4.02 in.
Bulk Pack
Black
51mm
Screw
Documents