missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Qorpak™ Glass Bottle Beakers with Green Thermoset F217 and PTFE Caps

Can be used for mixing, measuring and storage

Manufacturer:  Qorpak™ GLC01560

 View more versions of this product

Catalog No. 02992801


 

Description

Description

  • Designed with slight shoulder for easy pouring and handling
  • Molded graduations in mL and ounces
  • Made of clear glass
  • For use with biological and pathological specimens
  • For storage of industrial laboratory chemicals and reagents

Specifications

Specifications

16 oz., 480mL
16 oz.
F217 and PTFE
2.99 in.
Glass
Green
76mm
Screw
480mL
Thermoset
Clear
5.71 in.
24/Cs.
70-400
145mm
Documents