Qorpak™ Clear Straight Sided Round Bottles with Thermoset F217 and PTFE Caps, Cleaned for Volatiles
Offers maximum content visibility and integrity
$244.27 - $244.27
Specifications
Closure Liner F217 and PTFE
Closure Material Thermoset
Closure Type Screw
Closure Color Green
Color Clear
View More Specs
Products 1
Catalog Number Mfr. No. Capacity Diameter (English) Closure Size Height (English) Price Quantity  
Catalog Number Mfr. No. Capacity Diameter (English) Closure Size Height (English) Price Quantity  
02992785 View Documents Qorpak™
GLC01693
16 oz., 480 mL 3.58 in. 89-400 3.78 in. Case of 12 Each for $244.27
Description
Description
  • Without shoulders for maximum storage capacity
  • Green cap easily identifies PTFE liner system
  • Usage: analytical lab samples, high purity chemicals, strong acids solvents, environmental samples, pharmaceuticals, and diagnostic reagents
Certifications
Meets U.S. EPA Specifications and Guidelines for Contaminant-Free Sample Containers
Specifications
Specifications
F217 and PTFE
Screw
Clear
12/Cs.
Thermoset
Green
Glass