missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Qorpak™ Clear Standard Wide Mouth Bottles without Cap

Ideal general purpose bottle. Qorpak™ Clear Standard Wide Mouth Bottles without Cap provides maximum visibility and integrity of the sample contents.

$64.91 - $75.88

Specifications

Closure Type Screw Cap
Color Clear
Material Glass
View More Specs

Products 3
Catalog Number Mfr. No. Capacity Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Capacity Packaging Quantity Price Quantity  
02-993-030
View Documents Promotion Details
Qorpak™
GLA00869
4 oz., 120 mL Plain Carton 24/Cs.
Case of 24 Each for $64.91
 
02-993-118
View Documents Promotion Details
Qorpak™
GLA00882
64 oz., 1920 mL Conventional Pack 6/Cs.
Case of 6 Each for $75.88
 
02-993-096
View Documents Promotion Details
Qorpak™
GLA00883
128 oz., 3840 mL Conventional Pack 4/Cs.
Case of 4 Each for $69.03
 
Description

Description

Ideal general purpose bottle. Qorpak™ Clear Standard Wide Mouth Bottles without Cap provides maximum visibility and integrity of the sample contents.

  • Features wide opening
  • Applications include pharmaceutical, chemical, environmental, and food packaging and storing
Specifications

Specifications

Screw Cap
Clear
Glass
SDS
missing translation for 'documents'

missing translation for 'documents'