missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Qorpak™ Clear Standard Wide Mouth Bottles with White Polypropylene SturdeeSeal Cap

Ideal general purpose bottle. Qorpak™ Clear Standard Wide Mouth Bottles with White Polypropylene SturdeeSeal™ Cap provides maximum visibility and integrity of the sample contents.

$84.23 - $84.23

Specifications

Closure Liner SturdeeSeal™ Polyethylene Foam
Closure Material Polypropylene
Closure Type Screw
Closure Color White
Color Clear
View More Specs

Products 1
Catalog Number Mfr. No. Capacity (Metric) Quantity Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Quantity Price Quantity  
02-993-026 View Documents Qorpak™
GLC01758
120 mL 24/Cs. Case of 24 Each for $84.23
 
Description

Description

  • Features wide opening
  • Closures have sealing characteristics with good chemical resistant to acids, alcohols, alkalis, aqueous products, cosmetics, household oils and solvents
  • Applications include pharmaceutical, chemical, environmental, and food packaging and storing
Specifications

Specifications

SturdeeSeal™ Polyethylene Foam
Screw
Clear
Polypropylene
White
Glass