missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Qorpak™ Clear Standard Wide Mouth Bottles with Phenolic Solid PE Lined Cap

Provides maximum visibility and integrity of the sample contents. Qorpak™ Clear Standard Wide Mouth Bottles with Phenolic Solid PE Lined Cap are ideal for general purpose.

$100.40 - $100.40

Specifications

Closure Liner Polyethylene
Closure Material Phenolic
Closure Type Screw
Closure Color Black
Color Clear
View More Specs

Products 1
Catalog Number Mfr. No. Capacity Closure Size Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Capacity Closure Size Packaging Quantity Price Quantity  
02-992-835 View Documents Qorpak™
GLC01750
4 oz., 120 mL 48-400 Plain Carton 24/Cs. Case of 24 Each for $100.40
 
Description

Description

  • Features wide mouth opening
  • Closures offer good chemical resistance and are typically used with solids and powders
  • Resistant to hydro-carbons, alcohols, ketones and oils
  • Not for acids and alkalis
  • Applications include pharmaceutical, chemical, environmental and food packaging and storing
Specifications

Specifications

Polyethylene
Screw
Clear
Phenolic
Black
Glass