Qorpak™ Clear Standard Wide Mouth Bottles with Black Phenolic Pulp/Vinyl Lined Cap
Provides maximum visibility and integrity of the sample contents. Qorpak™ Clear Standard Wide Mouth Bottles with Black Phenolic Pulp/Vinyl Lined Cap are ideal for general purpose.
$98.00 - $147.63
Specifications
Closure Liner Pulp/Vinyl
Closure Material Phenolic
Closure Type Screw
Closure Color Black
Color Clear
View More Specs
Products 6
Catalog Number Mfr. No. Capacity Closure Size Height (Metric) Packaging Price Quantity  
Catalog Number Mfr. No. Capacity Closure Size Height (Metric) Packaging Price Quantity  
36-101-0481 View Documents Qorpak™
GLC01755
4 oz., 120 mL 48-400 86mm Carton Pack Case of 24 Each for $105.66
36-101-0482 View Documents Qorpak™
GLC01786
8 oz., 240 mL 58-400 109mm Carton Pack Case of 24 Each for $128.37
36-101-0377 View Documents Qorpak™
GLC01813*
16 oz., 480 mL 63-400 133mm Carton Pack Case of 24 Each for $147.63
02993145 View Documents Qorpak™
GLC01848
32 oz., 960 mL 70-400 177mm Carton Pack Case of 12 Each for $98.00
36-101-0380 View Documents Qorpak™
GLC01872
64 oz., 1920 mL 83-400 214mm Carton Pack Case of 6 Each for $101.48
36-101-0378 View Documents Qorpak™
GLC01894*
128 oz., 3840 mL 89-400 256mm Carton Pack Case of 4 Each for $98.40
Description
Description
  • Features wide mouth opening
  • Used for food, beverage, medical and chemical applications packed at less than 120°F, including mild acids, alkalis, solvents, alcohols, oils and aqueous products
  • Not recommended for active hydrocarbons or bleaches
Specifications
Specifications
Pulp/Vinyl
Screw
Clear
Phenolic
Black
Glass