missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Qorpak™ Clear PVC Cylinder Bottles with White Polypropylene SturdeeSeal™ PE Foam Lined Cap

For handling liquid samples. Qorpak™ Clear PVC Cylinder Bottles with White Polypropylene SturdeeSeal™ PE Foam Lined Cap are transparent and helps for easy visual identification of contents inside.

Manufacturer:  Qorpak™ PLC05772

 View more versions of this product

Catalog No. 02-993-355


 

Description

Description

  • Features narrow mouth
  • Shoulder style allows pouring of small quantities without spilling
Specifications

Specifications

8 oz., 240mL
8 oz.
SturdeeSeal™ Polyethylene Foam
1.93 in.
PVC
White
49mm
24-410
240mL
Polypropylene
Clear
6.34 in.
360/CS
24-410
161mm
Screw
Documents