Qorpak™ Clear PET Packer Bottles, Precleaned — Ultra Clean
Precleaned for elimination of contaminants. Qorpak™ Clear PET Packer Bottles, Precleaned — Ultra Clean feature a proprietary cleaning process that removes up to 99.5% of particles that are 5 microns and larger.
$488.94 - $488.94
Specifications
Closure Liner SturdeeSeal™ Polyethylene Foam
Closure Material Polypropylene
Closure Color White
Closure Type Screw
Color Clear
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Capacity Closure Size Quantity Material Price Quantity  
Catalog Number Mfr. No. Capacity Closure Size Quantity Material Price Quantity  
02-991-839 Qorpak™
PLC06806
17 oz., 500mL 45-400 24/Cs. PETE Case of 24 Each for $488.94
Specifications
Description
Description & Specifications
Specifications
SturdeeSeal™ Polyethylene Foam
White
Clear
Polypropylene
Screw
PETE