Please login to your online account to display your discounted pricing

Qorpak™ Clear Graduated Medium Round Ultra Clean Bottles


For use with biological and pathological specimens. Qorpak™ Clear Graduated Medium Round Ultra Clean Bottles are used for storing industrial laboratory chemicals and reagents.

Manufacturer: qorpak™  GLC01562

 View more versions of this product

Catalog No. 02-993-106

  • / Case of 24 Each
 

Description & Specifications

Specifications

Capacity 16 oz., 480mL
Capacity (Metric) 480mL
Capacity (English) 16 oz.
Closure Material Thermoset
Closure Liner PTFE
Color Clear
Diameter (English) 2.99 in.
Height (English) 5.71 in.
Material Glass
Quantity 24/Cs.
Closure Color Green
Closure Size 70-400
Diameter (Metric) 76mm
Height (Metric) 145mm
Closure Type Screw

  • Features wide mouth
  • Slight shoulder for easy pouring and handling
  • Available with caps made of different materials (Green thermoset F217 with PTFE and black phenolic pulp/vinyl)