missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Qorpak™ Clear Graduated Medium Round Ultra Clean Bottles with Phenolic Caps

For use with biological and pathological specimens. Qorpak™ Clear Graduated Medium Round Ultra Clean Bottles are used for storing industrial laboratory chemicals and reagents.

Manufacturer:  Qorpak™ GLC01467

 View more versions of this product

Catalog No. 02-992-375



Description

Description

  • Features wide mouth
  • Slight shoulder for easy pouring and handling
Specifications

Specifications

1 oz., 30mL
1 oz.
Pulp/Vinyl
1.34 in.
Glass
Black
34mm
Screw
30mL
Phenolic
Clear
2.68 in.
48/Cs.
33-400
68mm
Documents