Please login to your online account to display your discounted pricing

Qorpak™ Clear Graduated Medium Round Ultra Clean Bottles


For use with biological and pathological specimens. Qorpak™ Clear Graduated Medium Round Ultra Clean Bottles are used for storing industrial laboratory chemicals and reagents.

Manufacturer: qorpak™  GLC01452

 View more versions of this product

Catalog No. 02-992-373

  • / Case of 48 Each
 

Description & Specifications

Specifications

Capacity (Metric) 30mL
Capacity (English) 1 oz.
Closure Color Green
Closure Screw
Closure Material Thermoset
Closure Liner PTFE
Closure Size 33-400
Color Clear
Diameter (English) 1.34 in.
Diameter (Metric) 34mm
Height (Metric) 68mm
Material Glass
Quantity 48/Cs.
Height (English) 2.68 in.
Capacity 1 oz., 30mL

  • Features wide mouth
  • Slight shoulder for easy pouring and handling
  • Available with caps made of different materials (Green thermoset F217 with PTFE and black phenolic pulp/vinyl)