Please login to your online account to display your discounted pricing

Qorpak™ Clear Graduated Medium Round Ultra Clean Bottles


For use with biological and pathological specimens. Qorpak™ Clear Graduated Medium Round Ultra Clean Bottles are used for storing industrial laboratory chemicals and reagents.

Manufacturer: qorpak™  GLC01524

 View more versions of this product

Catalog No. 02-992-372

  • / Case of 24 Each
 

Description & Specifications

Specifications

Closure Color Black
Closure Type Screw
Height (Metric) 102mm
Diameter (Metric) 51mm
Closure Size 48-400
Diameter (English) 2.01 in.
Material Glass
Closure Material Phenolic
Capacity (English) 4 oz.
Quantity 24/Cs.
Color Clear
Capacity 4 oz., 120mL
Height (English) 4.02 in.
Closure Liner Pulp/Vinyl
Capacity (Metric) 120mL

  • Features wide mouth
  • Slight shoulder for easy pouring and handling
  • Available with caps made of different materials (Green thermoset F217 with PTFE and black phenolic pulp/vinyl)