Please login to your online account to display your discounted pricing

Qorpak™ Clear Graduated Medium Round Ultra Clean Bottles


For use with biological and pathological specimens. Qorpak™ Clear Graduated Medium Round Ultra Clean Bottles are used for storing industrial laboratory chemicals and reagents.

Manufacturer: qorpak™  GLC01576

 View more versions of this product

Catalog No. 02-992-369

  • / Case of 24 Each
 

Description & Specifications

Specifications

Capacity (Metric) 480mL
Capacity (English) 16 oz.
Closure Color Black
Closure Screw
Closure Material Phenolic
Closure Liner Pulp/Vinyl
Closure Size 70-400
Color Clear
Diameter (English) 2.99 in.
Diameter (Metric) 76mm
Height (English) 5.71 in.
Height (Metric) 145mm
Material Glass
Quantity 24/Cs.
Capacity 16 oz., 480mL

  • Features wide mouth
  • Slight shoulder for easy pouring and handling
  • Available with caps made of different materials (Green thermoset F217 with PTFE and black phenolic pulp/vinyl)