Please login to your online account to display your discounted pricing

Qorpak™ Clear Graduated Medium Round Ultra Clean Bottles


For use with biological and pathological specimens. Qorpak™ Clear Graduated Medium Round Ultra Clean Bottles are used for storing industrial laboratory chemicals and reagents.

Manufacturer: qorpak™  GLC01534

 View more versions of this product

Catalog No. 02-992-367

  • / Case of 24 Each
 

Description & Specifications

Specifications

Closure Color Green
Closure Size 58-400
Height (Metric) 127mm
Diameter (Metric) 62mm
Closure Type Screw
Capacity (English) 8 oz.
Diameter (English) 2.44 in.
Quantity 24/Cs.
Material Glass
Color Clear
Capacity (Metric) 240mL
Closure Liner PTFE
Capacity 8 oz., 240mL
Height (English) 5 in.
Closure Material Thermoset

  • Features wide mouth
  • Slight shoulder for easy pouring and handling
  • Available with caps made of different materials (Green thermoset F217 with PTFE and black phenolic pulp/vinyl)