Please login to your online account to display your discounted pricing

Qorpak™ Clear Graduated Medium Round Ultra Clean Bottles


For use with biological and pathological specimens. Qorpak™ Clear Graduated Medium Round Ultra Clean Bottles are used for storing industrial laboratory chemicals and reagents.

Manufacturer: qorpak™  GLC01553

 View more versions of this product

Catalog No. 02-992-365

  • / Case of 24 Each
 

Description & Specifications

Specifications

Closure Color Black
Closure Type Screw
Closure Size 58-400
Diameter (Metric) 62mm
Height (Metric) 127mm
Diameter (English) 2.44 in.
Material Glass
Capacity (English) 8 oz.
Color Clear
Quantity 24/Cs.
Capacity 8 oz., 240mL
Closure Liner Pulp/Vinyl
Capacity (Metric) 240mL
Height (English) 5 in.
Closure Material Phenolic

Vacuum and Ionization Cleaning Process: Compressed air passes through a condensate separator and coalescing filter to remove aerosols and fine dust particles. Next, the air passes through an absorbent filter to remove gases. A vacuum draws the particulate-laden atmosphere out of the container. Individually cleaned closures are immediately placed on the container. Contaminants such as loose dirt, carton lint, aerosols, and fine glass particles are removed by the process.
  • Clear glass provides maximum content visibility and minimal chemical inference with bottle's contents
  • Attached caps reduce carton dust