Qorpak™ Clear Boston Round Bottles, Vacuum and Ionized with Thermoset F217 and PTFE Cap
General use bottles ideally used when content integrity and visibility is important. Qorpak™ Clear Boston Round Bottles, Vacuum and Ionized with Thermoset F217 and PTFE Cap is cleaned using Vacuum and Ionization process.
$97.00 - $167.83
Specifications
Cleaning Procedure Vacuum and ionized
Closure Liner F217 and PTFE
Closure Material Thermoset
Closure Type Screw
Closure Color Green
View More Specs
Products 3
Catalog Number Mfr. No. Capacity Packaging Price Quantity  
Catalog Number Mfr. No. Capacity Packaging Price Quantity  
03-328-1A View Documents Qorpak™
GLC01085
1 oz., 30mL Convenience Pack Case of 48 Each for $167.83
02-992-192 View Documents Qorpak™
GLC01102
2 oz., 60 mL Convenience Pack Case of 24 Each for $97.00
02-992-205 View Documents Qorpak™
GLC01126*
4 oz., 120 mL Convenience Pack Case of 24 Each for $109.74
Description
Description
Vacuum and Ionization Cleaning Process: Compressed air passes through a condensate separator and coalescing filter to remove aerosols and fine dust particles. Next, the air passes through an absorbent filter to remove gases. A vacuum draws the particulate-laden atmosphere out of the container. Individually cleaned closures are immediately placed on the container. Contaminants such as loose dirt, carton lint, aerosols, and fine glass particles are removed by the process.
Clear Boston Round Bottles
  • Feature round body, curved shoulders, and narrow screw neck opening
  • Provides a clean final appearance
  • Air inside bottle does not interfere with product, rendering them odor-free and tasteless
Specifications
Specifications
Vacuum and ionized
Thermoset
Green
Glass
F217 and PTFE
Screw
Clear