Qorpak™ Clear Boston Round Bottles with Polypropylene Cap and PTFE Disc
For liquids, product storage, and field or plant sampling projects. Qorpak™ Clear Boston Round Bottles with Polypropylene Cap and PTFE Disc help to insure product integrity for long term storage.
$108.06 - $108.06
Specifications
Closure Material Polypropylene
Closure Type Screw
Closure Color Black
Color Clear
Material Glass
View More Specs
Products 1
Catalog Number Mfr. No. Capacity Closure Size Height (English) Quantity Price Quantity  
Catalog Number Mfr. No. Capacity Closure Size Height (English) Quantity Price Quantity  
02-992-191 View Documents Qorpak™
GLC01209
32 oz., 960 mL 33-400 8.2 in. 12/Cs. (Plain Ctn) Case of 12 Each for $108.06
Description
Description
  • Features a narrow screw neck opening, curved shoulders, and a round body
  • Glass offers minimal chemical interference with contents
  • Environmental sensitive, helps eliminate waste
Specifications
Specifications
Polypropylene
Black
Glass
Screw
Clear