Qorpak™ Clear Boston Round Bottles with Black Phenolic PolyCone Caps
General-use bottles for liquids, product storage, and field or plant sampling projects. Qorpak™ Clear Boston Round Bottles with Black Phenolic PolyCone Cap offer maximum visibility and sample integrity.
$76.43 - $308.54
Specifications
Closure Liner Polycone
Closure Material Phenolic
Closure Type Screw
Closure Color Black
Color Clear
View More Specs
Products 4
Catalog Number Mfr. No. Capacity Closure Size Height (English) Quantity Price Quantity  
Catalog Number Mfr. No. Capacity Closure Size Height (English) Quantity Price Quantity  
02991945 View Documents Qorpak™
GLC01193
16 oz., 480 mL 28-400 6.6 in. 60/Cs. (Bulk Pk.) Case of 60 Each for $308.54
02992208 View Documents Qorpak™
GLC01196
16 oz., 480 mL 28-400 6.6 in. 12/Cs. (Conv. Pk) Case of 12 Each for $76.43
02-992-186 View Documents Qorpak™
GLC01228*
32 oz., 960 mL 33-400 8.2 in. 12/Cs. (Qorpak Ctn) Case of 12 Each for $108.63
02-992-895 View Documents Qorpak™
GLC01226
32 oz., 960 mL 33-400 8.2 in. 12/Cs. (Plain Ctn) Case of 12 Each for $102.12
Description
Description
  • Features a round body, rounded shoulders and narrow screw neck opening
  • Environmental sensitive help eliminate waste and ensure product integrity for long term storage
  • Form tight seal and offer chemical barrier
  • Liner molds itself around the sealing areas
Specifications
Specifications
Polycone
Screw
Clear
Phenolic
Black
Glass