Qorpak™ Black Phenolic Rubber Lined Caps
Intented for repeated use. Qorpak™ Black Phenolic Rubber Lined Caps are autoclavable.
Supplier: Qorpak™ CAP05781
Description
- Liner exhibit resiliency
- Resistant to moisture vapor
- Provides consistent adherence to close dimensional tolerance
- Available in ribbed and smooth style
- Available as bulk/bagged packaging
Specifications
Smooth | |
Wide mouth bottle | |
Phenolic Resin | |
Bulk | |
FDA 21 CFR 177.2600 |
Rubber | |
Black | |
1400 Cs. | |
48-400 |