Please login to your online account to display your discounted pricing

Qorpak™ Black Phenolic Pulp/Aluminum Foil Lined Caps


Ideal for non-acid and non-alkaline products. Qorpak™ Black Phenolic Pulp/Aluminum Foil Lined Caps are typically used for food applications.

Manufacturer: qorpak™  CAP00312

 View more versions of this product

Catalog No. 02-993-241

  • / Case of 144 Each
 

Description & Specifications

Specifications

Closure Liner Pulp/Aluminum Foil
Closure Type Smooth
Color Black
For Use With Wide mouth bottle
Neck Finish 70-400
No. per Case 144 Cs.
No. per Pack 12/Pk.
Packaging Bagged

  • Closure liners free of odor and taste
  • Used for organic solvents, chrome cleaners, brake fluids and mineral oils
  • Neck finish for narrow bottles is 425 GPI and 400 for wider
  • Available as ribbed style
  • Available in bulk/bagged packaging