Please login to your online account to display your discounted pricing

Qorpak™ Black Phenolic 0.030 Solid Polyethylene Lined Caps


Ideal for nail polish remover and higher concentration acids and alkalis. Qorpak™ Black Phenolic 0.030 Solid Polyethylene Lined Caps offer good chemical resistance and low moisture vapor transmission rate.

Manufacturer: qorpak™  CAP00207

 View more versions of this product

Catalog No. 02-991-206

  • / Case of 2400 Each
 

Description & Specifications

Specifications

Closure Type Smooth
Neck Finish 33-400
For Use With (Equipment) Wide mouth bottle
No. per Case 2400
Color Black
Packaging Bulk
Closure Liner Polyethylene

  • Paperless composition avoids contamination from fiber or paper particles
  • For non-oil products filled at room temperature