Qorpak™ Amber Wide Mouth Packer Bottles with Black Phenolic PolyCone Cap
General use bottles ideal for UV sensitive contents. Qorpak™ Amber Wide Mouth Packer Bottles with Black Phenolic PolyCone Cap are used in chemical and environmental applications.
$89.63 - $89.63
Specifications
Closure Liner Polycone
Closure Material Phenolic
Closure Type Screw
Closure Color Black
Color Amber
View More Specs
Products 1
Catalog Number Mfr. No. Capacity Closure Size Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Capacity Closure Size Packaging Quantity Price Quantity  
02-993-029 View Documents Qorpak™
GLC02101
2 oz., 60 mL 33-400 Conventional pack 24/Cs. Case of 24 Each for $89.63
Description
Description
  • Feature a wide opening for making them the ideal general purpose bottle for solids
  • Classic "vitamin bottles" and are often used in pharmaceutical packaging
Specifications
Specifications
Polycone
Screw
Amber
Phenolic
Black
Glass