Please login to your online account to display your discounted pricing
Qorpak™ Amber Wide Mouth Packer Bottles, Vacuum and Ionized with Phenolic PolyCone Cap
Provides additional protection for light-sensitive contents by blocking UV rays. Qorpak™ Amber Wide Mouth Packer Bottles, Vacuum and Ionized with Phenolic PolyCone Cap is useful for wide range of applications including pharmaceutical, chemical and environmental.
$110.37 - $762.46
Specifications
Cleaning Procedure Vacuum and ionized
Closure Liner Polycone
Closure Material Phenolic
Closure Screw
Closure Color Black
View More Specs
Products
Catalog Number Mfr. No. Capacity Packaging Type Quantity Closure Size Price Quantity    
02-991-854
qorpak™
GLC07989
2.02 oz., 60mL Conventional pack 24/Cs. 33-400 Case of 24 Each for $110.37
02-991-934
qorpak™
GLC06345
4 oz., 120mL Bulk pack 180/Cs. 38-400 Case for $762.46
Description & Specifications
Specifications
Cleaning Procedure Vacuum and ionized
Closure Liner Polycone
Closure Material Phenolic
Closure Screw
Closure Color Black
Color Amber
Material Glass
  • Air inside the bottle is odor-free, tasteless and does not react with product
  • Separate washing not required, thereby saving time
  • Final finished product kept clean and dust-free
  • Contaminants such as loose dirt, carton lint, aerosols, and fine glass particles are removed by the Vacuum and Ionization process