Please login to your online account to display your discounted pricing

Qorpak™ Amber and Clear Borosilicate Compound Vials: Without Cap

Wide mouth facilitates easy sampling

$205.34 - $205.34

Specifications

Closure Type Screw cap
Diameter (English) Outside 0.59 in.
Diameter (Metric) Outside 15mm
Color Clear
Material Borosilicate Glass
View More Specs
Products
Catalog Number Mfr. No. Capacity (Metric) Height (Metric) Quantity Color Price Quantity    

02-991-622

 
qorpak™
GLA07794
0.95mL 19mm 144/Cs. Clear Case of 144 Each for $205.34
Description & Specifications

Specifications

Closure Type Screw cap
Diameter (English) Outside 0.59 in.
Diameter (Metric) Outside 15mm
Color Clear
Material Borosilicate Glass
Type Compound Vial
For Use With (Application) Wide mouth facilitates easy sampling
Thread Style Screw

  • Low profile vials for better stability
  • Vial only
  • Available in amber or clear colors