Qorpak™ Amber Boston Round Bottles, Vacuum and Ionized with Thermoset F217 and PTFE Cap
General use bottles ideal for UV sensitive contents. Qorpak™ Amber Boston Round Bottles, Vacuum and Ionized: With Thermoset F217 and PTFE Cap ensures product integrity for long-term storage.
$132.29 - $135.66
Specifications
Closure Liner F217 and PTFE
Closure Material Thermoset
Closure Color Green
Closure Type Screw
Color Amber
View More Specs
Products 2
Catalog Number Mfr. No. Capacity Closure Size Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Capacity Closure Size Packaging Quantity Price Quantity  
02-992-721 View Documents Qorpak™
GLC01950
8 oz., 240 mL 24-400 Convention pack 24/Cs. Case of 24 Each for $132.29
36-101-1036 View Documents Qorpak™
GLC02006
32 oz., 960 mL 33-400 Convention pack 12/Cs. Case of 12 Each for $135.66
Description
Description
Vacuum and Ionization Cleaning Process: Compressed air passes through a condensate separator and coalescing filter to remove aerosols and fine dust particles. Next, the air passes through an absorbent filter to remove gases. A vacuum draws the particulate-laden atmosphere out of the container. Individually cleaned closures are immediately placed on the container. Contaminants such as loose dirt, carton lint, aerosols, and fine glass particles are removed by the process.
Amber Boston Round Bottles
  • Narrow screw neck opening with curved shoulders
  • Glass offers minimal chemical interference with contents
Specifications
Specifications
F217 and PTFE
Green
Amber
Thermoset
Screw
Glass