Qorpak™ Amber Boston Round Bottles with White Polypropylene Hole Cap and Bonded PTFE/Silicone Septa
General use bottles ideal for UV sensitive contents. Qorpak™ Amber Boston Round Bottles with White Polypropylene Hole Cap and Bonded PTFE/Silicone Septa ensures product integrity for long-term storage.
$121.23 - $121.23
Specifications
Closure Material Polypropylene
Closure Type Screw
Closure Color White
Color Amber
Material Glass
View More Specs
Products 1
Catalog Number Mfr. No. Capacity Height (Metric) Height (English) Quantity Price Quantity  
Catalog Number Mfr. No. Capacity Height (Metric) Height (English) Quantity Price Quantity  
02-992-400 View Documents Qorpak™
GLC06424
16 oz., 480 mL 168mm 6.6 in. 12/Cs. Case of 12 Each for $121.23
Description
Description
  • Features a narrow screw neck opening, curved shoulders, and a round body
  • Glass offers minimal chemical interference with contents
  • Amber color absorb all wavelengths in the ultraviolet spectrum
Specifications
Specifications
Polypropylene
White
Glass
Screw
Amber