Qorpak™ Amber Boston Round Bottles with Green Thermoset F217 and PTFE Cap
General use bottles ideal for UV sensitive contents. Qorpak™ Amber Boston Round Bottles with Green Thermoset F217 and PTFE Cap ensures product integrity for long-term storage.
$107.76 - $273.14
Specifications
Closure Liner F217 and PTFE
Closure Material Thermoset
Closure Type Screw
Closure Color Green
Color Amber
View More Specs
Products 2
Catalog Number Mfr. No. Capacity Diameter (English) Closure Size Height (English) Price Quantity  
Catalog Number Mfr. No. Capacity Diameter (English) Closure Size Height (English) Price Quantity  
02-993-039 View Documents Qorpak™
GLC01926
4 oz., 120 mL 1.88 in. 22-400 4.4 in. Case of 24 Each for $107.76
02-992-815 View Documents Qorpak™
GLC02001
32 oz., 960 mL 3.7 in. 33-400 8.2 in. Case of 30 Each for $273.14
Description
Description
  • Features a narrow screw neck opening, curved shoulders, and a round body
  • Glass offers minimal chemical interference with contents
  • Amber color absorb all wavelengths in the ultraviolet spectrum
Specifications
Specifications
F217 and PTFE
Screw
Amber
Thermoset
Green
Glass