Please login to your online account to display your discounted pricing
Qorpak™ Amber Boston Round Bottles with Black Phenolic Rubber Cap
General use bottles ideal for UV sensitive contents. Qorpak™ Amber Boston Round Bottles with Black Phenolic Rubber Cap ensures product integrity for long-term storage.
$96.57 - $131.29
Specifications
Closure Material Phenolic
Closure Color Black
Closure Type Screw
Color Amber
Material Glass
View More Specs
Products
Catalog Number Mfr. No. Capacity Packaging Type Quantity Closure Size Price Quantity    
02-992-484
qorpak™
GLC01981
16 oz., 480mL Case of 12 Each for $96.57
02-993-078
qorpak™
GLC01959
8 oz., 240mL Case of 24 Each for $131.29
02-993-112
qorpak™
GLC01930
4 oz., 120mL Case of 24 Each for $105.46
Description & Specifications
Specifications
Closure Material Phenolic
Closure Color Black
Closure Type Screw
Color Amber
Material Glass
  • Features a narrow screw neck opening, curved shoulders, and a round body
  • Glass offers minimal chemical interference with contents
  • Amber color absorb all wavelengths in the ultraviolet spectrum