Qorpak™ Amber Boston Round Bottles with Black Phenolic PolyCone Cap
General use bottles ideal for UV sensitive contents. Qorpak™ Amber Boston Round Bottles with Black Phenolic PolyCone Cap ensures product integrity for long-term storage.
$76.34 - $1,083.63
Specifications
Closure Liner Polycone
Closure Material Phenolic
Closure Type Screw
Closure Color Black
Color Amber
View More Specs
Products 8
Catalog Number Mfr. No. Capacity Height (Metric) Quantity Price Quantity  
Catalog Number Mfr. No. Capacity Height (Metric) Quantity Price Quantity  
02991993 View Documents Qorpak™
GLC01898
1 oz., 30mL 79mm 360/Cs. Case of 432 Each for $1,083.63
02993138 View Documents Qorpak™
GLC01935
4 oz., 120 mL 112mm 24/Cs. Case of 24 Each for $94.86
02992141 View Documents Qorpak™
GLC01932
4 oz., 120 mL 112mm 128/Cs. Case of 128 Each for $416.57
02991866 View Documents Qorpak™
GLC01960
8 oz., 240 mL 138mm 96/Cs. Case of 108 Each for $422.17
02993143 View Documents Qorpak™
GLC01961
8 oz., 240 mL 138mm 24/Cs. Case of 24 Each for $111.97
02991848 View Documents Qorpak™
GLC01982
16 oz., 480 mL 168mm 60/Cs. Case of 60 Each for $311.09
02993117 View Documents Qorpak™
GLC01985
16 oz., 480 mL 168mm 12/Cs. Case of 12 Each for $76.34
02-992-476 View Documents Qorpak™
GLC02012*
32 oz., 960 mL 210mm 12/Cs. Case of 12 Each for $112.66
Description
Description
  • Features a narrow screw neck opening, curved shoulders, and a round body
  • Glass offers minimal chemical interference with contents
  • Amber color absorb all wavelengths in the ultraviolet spectrum
Specifications
Specifications
Polycone
Screw
Amber
Phenolic
Black
Glass