missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Qorpak™ Amber Borosilicate Sample Vials: Without Caps

Offers protection from UV rays and harmful light for light sensitive products

Manufacturer:  Qorpak™ GLA00801

 View more versions of this product

Catalog No. 02-991-507


 

Specifications Description & Specifications

Specifications

Screw cap
5 dr., 20mL
27.75mm
5 dr.
Sample Vial
57mm
Used for general purposes, protects against harmful UV rays
5 dr.
20mL
1.09 in.
Amber
Borosilicate Glass
20mL
2.24 in.
Screw
144/Cs.