Please login to your online account to display your discounted pricing

Qorpak™ Amber Borosilicate Sample Vials: With Phenolic PolyCone Cap

Offers protection from UV rays and harmful light for light sensitive products

$495.74 - $615.49

Specifications

Closure Type Screw cap
Diameter (Metric) Outside 27.75mm
Color Amber
Material Borosilicate Glass
Type Sample Vial
View More Specs
Products
Catalog Number Mfr. No. Capacity (Metric) Height (Metric) Quantity Price Quantity    

02-991-758

 
qorpak™
GLC01061
20mL 57mm 144/Cs. Case of 144 Each for $495.74

02-992-609

 
qorpak™
GLC04892
40mL 95mm 144/Cs. Case of 144 Each for $615.49
Description & Specifications

Specifications

Closure Type Screw cap
Diameter (Metric) Outside 27.75mm
Color Amber
Material Borosilicate Glass
Type Sample Vial
For Use With (Application) Used for general purposes, protects against harmful UV rays
Material Closure Phenolic
Material (Liner) Polycone
Thread Style Screw

  • Used for general purposes
  • Capacity range: 0.50 to 10 dr. (1.85 to 40mL)