missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Qorpak™ Alpha Caps for F-Style Cans

Odor-free and tasteless nature make it more feasible for food applications and non-acid, non-alkaline products. Qorpak™ Alpha Caps for F-Style Cans are made of metal.

$1,787.33 - $1,787.33

Specifications

Color Silver
Material Metal
For Use With (Equipment) Vegetable and mineral oil
No. per Case 2700
View More Specs

Products 1
Catalog Number Mfr. No. Closure Liner Price Quantity  
Catalog Number Mfr. No. Closure Liner Price Quantity  
02-991-311 View Documents Qorpak™
MET05430
Solvseal Case of 2700 Each for $1,787.33
 
Description

Description

  • Impact resistant and shows wide range of temperature tolerances
  • Available with pulp/alum foil or Solvseal liner
  • Solvseal liners are ideal for vegetable and mineral oils, weak acids and alkalis, alcohols and organic solvents
  • Aluminum foil lined caps are used for food applications and can be used for non-acid, non-alkaline products
Specifications

Specifications

Silver
Metal
Vegetable and mineral oil
2700