Qorpak™ White Polypropylene SturdeeSeal PE Foam Lined Caps
Features excellent sealing properties
$344.30 - $1,070.89
Specifications
Closure Liner SturdeeSeal™ Polyethylene Foam
Closure Type Ribbed
Color White
View More Specs
Products 6
Catalog Number Mfr. No. Neck Finish For Use With (Equipment) No. per Case Packaging Price Quantity  
Catalog Number Mfr. No. Neck Finish For Use With (Equipment) No. per Case Packaging Price Quantity  
02-991-068 View Documents Qorpak™
CAP00324
89-400 Wide mouth bottle 608 Cs. Bulk Case of 624 Each for $933.39
02-991-271 View Documents Qorpak™
CAP00359
33-400 Wide mouth bottle 5000 Cs. Bulk Case of 5000 Each for $1,030.61
02-991-343 View Documents Qorpak™
CAP00389
48-400 Wide mouth bottle 2500 Cs. Bulk Case of 2500 Each for $901.73
02-991-410 View Documents Qorpak™
CAP05103
89-400 Wide mouth bottle 144 Cs. Bagged Case of 144 Each for $365.61
02-991-440 View Documents Qorpak™
CAP05785
20-410 Bottle 7200 Cs. Bulk Case of 7200 Each for $1,070.89
02-991-541 View Documents Qorpak™
CAP06623
33-400 Wide mouth bottle 576 Cs. Bagged Case of 576 Each for $344.30
Description
Description
  • Can be used when good impact strength and pliability are required
  • Resistant to many chemicals including acids, alcohols, alkalis, aqueous products, cosmetics, household oils and solvents
  • Available in ribbed style
  • Available in bulk or bagged packing
Specifications
Specifications
SturdeeSeal™ Polyethylene Foam
Ribbed
White