Qorpak™ White Polypropylene SturdeeSeal PE Foam Lined Caps
Features excellent sealing properties
Supplier: Qorpak™ CAP05103
Description
- Can be used when good impact strength and pliability are required
- Resistant to many chemicals including acids, alcohols, alkalis, aqueous products, cosmetics, household oils and solvents
- Available in ribbed style
- Available in bulk or bagged packing
Specifications
Ribbed | |
Wide mouth bottle | |
Polypropylene | |
12/Pk. | |
89-400 |
Polyethylene Foam | |
White | |
144 Cs. | |
Bagged |