Please login to your online account to display your discounted pricing

Qorpak™ White Polypropylene SturdeeSeal PE Foam Lined Caps


Features excellent sealing properties

Manufacturer: qorpak™  CAP00389

 View more versions of this product

Catalog No. 02-991-343

  • / Case of 2500 Each
 

Description & Specifications

Specifications

Closure Liner SturdeeSeal™ Polyethylene Foam
Color White
No. per Case 2500 Cs.
Packaging Bulk
For Use With (Equipment) Wide mouth bottle
Neck Finish 48-400
Closure Type Ribbed