missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ QNZ, 98%

Manufacturer:  Alfa Aesar™ J67487EXD

 View more versions of this product

Catalog No. AAJ67487EXD

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

356.43
Solid
QNZ, 98%
6-Amino-4-[2-(4-phenoxyphenyl)ethylamino]quinazoline;
C22H20N4O
-30°C to -10°C
545380-34-5
98%
White
0.5mg