Please login to your online account to display your discounted pricing

Tocris™ Qc 1

Threonine dehydrogenase inhibitor

$214.12 - $895.76

Specifications

Product Type Qc 1
Inhibitors Qc 1
Molecular Weight 455.45
Purity >98
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

505510R

 
tocris™
505510
10mg Each for $214.12

505550R

 
tocris™
505550
50mg Each for $895.76
Description & Specifications

Specifications

Product Type Qc 1
Inhibitors Qc 1
Molecular Weight 455.45
Purity >98

Reversible, non-competitive inhibitor of threonine dehydrogenase (TDH) (IC50 ∽500nM). Induces autophagy and inhibits cell proliferation in mouse embryonic stem (ES) cells.

  • Chemical Name: 1,2,3,4-Tetrahydro-4-oxo-N-(phenylmethyl)-2-thioxo-3-[3-(trifluoromethyl)phenyl]-7-quinazolinecarboxamide
  • Formula: C23H16F3N3O2S
  • CAS Number: 403718-45-6
  • Solubility: Soluble to 50mM in DMSO
  • Purity: >98