Please login to your online account to display your discounted pricing

Qa-2 Mouse anti-Mouse, FITC, Clone: 69H1-9-9, eBioscience™

Mouse Monoclonal Antibody

$114.55 - $404.55

Specifications

Antigen Qa-2
Clone 69H1-9-9
Host Species Mouse
Gene Alias Qa2
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

509959

 
life technologies
LS11599681
50μg Each for $114.55

509960

 
life technologies
LS11599682
100μg Each for $187.05

509961

 
life technologies
LS11599685
500μg Each for $404.55
Description & Specifications

Specifications

Antigen Qa-2
Clone 69H1-9-9
Host Species Mouse
Gene Alias Qa2
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate FITC
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The 69H1-9-9 monoclonal antibody reacts with the mouse Qa-2 molecule, a GPI-linked MHC class Iβ surface molecule. Qa-2 is expressed by mature T and B cells, and its expression level is different depending on the laboratory mouse strain.