missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Q94 hydrochloride, Tocris™

Negative allosteric modulator at PAR1 receptor

$176.10 - $741.11

Chemical Identifiers

Assay Percent Range >99%
Molecular Formula C21H17ClN2·HCl
Formula Weight 369.29
Synonym 1-[(4-Chlorophenyl)methyl]-2-(phenylmethyl)-1H-benzimidazole hydrochloride
Chemical Name or Material Q94 hydrochloride
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
475550 R & D Systems
475550
50mg Each for $741.11
475510 R & D Systems
475510
10mg Each for $176.10
Specifications Description & Specifications

Chemical Identifiers

>99%
369.29
Q94 hydrochloride
C21H17ClN2·HCl
1-[(4-Chlorophenyl)methyl]-2-(phenylmethyl)-1H-benzimidazole hydrochloride

Specifications

50mg
>99%
369.29
Q94 hydrochloride
1052076-77-3
C21H17ClN2·HCl
1-[(4-Chlorophenyl)methyl]-2-(phenylmethyl)-1H-benzimidazole hydrochloride

Safety and Handling

Recommended Storage : Store at +4°C