Please login to your online account to display your discounted pricing

Pyruvic aldehyde dimethyl acetal, 98%, ACROS Organics™


Quantity 5g

Manufacturer: thermo fisher scientific  173800050

 View more versions of this product

Catalog No. AC173800050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 118.13
Physical Form Liquid
Quantity 5g
Color Clear colorless to yellow
Density 1g/mL
EINECS Number 228-735-4
TSCA TSCA
Beilstein 01, IV, 3631
Synonym 1, 1-Dimethoxyacetone
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Flammable liquid and vapor.
Refractive Index 1.3970 to 1.3990 (20°C, 589nm)
Health Hazard 3 GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 GHS Signal Word: Warning
Chemical Name or Material Pyruvic aldehyde dimethyl acetal
Linear Formula CH3COCH(OCH3)2
Molecular Formula C5H10O3
MDL Number MFCD00008758
Name Note 98%
Assay Percent Range 98%
CAS 6342-56-9

Specifications

Chemical Name or Material Pyruvic aldehyde dimethyl acetal
Name Note 98%
CAS 6342-56-9
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C5H10O3
Linear Formula CH3COCH(OCH3)2
Formula Weight 118.13
MDL Number MFCD00008758
Synonym 1, 1-Dimethoxyacetone
Accessories

GHS Signal Word: Warning

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Flammable liquid and vapor.

SDS & Safety Documents