missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Pyruvic Acid | |
| 13°C | |
| 62°C | |
| MFCD00002585 | |
| 3265 | |
| LCTONWCANYUPML-UHFFFAOYSA-N | |
| 2-oxopropanoic acid | |
| 1060 | |
| 88.06 | |
| Liquid |
| 127-17-3 | |
| Yellow | |
| C3H4O3 | |
| 25 g | |
| pyruvic acid, pyroracemic acid, 2-oxopropionic acid, acetylformic acid, alpha-ketopropionic acid, 2-ketopropionic acid, 2-oxopropanoate, propanoic acid, 2-oxo, 2-oxo-propionic acid, 2-oxopropansaeure | |
| CC(=O)C(O)=O | |
| 88.06 | |
| CHEBI:32816 | |
| ≥97.0% (GC) |
Chemical Identifiers
| 127-17-3 | |
| 88.06 | |
| LCTONWCANYUPML-UHFFFAOYSA-N | |
| 1060 | |
| 2-oxopropanoic acid |
| C3H4O3 | |
| MFCD00002585 | |
| pyruvic acid, pyroracemic acid, 2-oxopropionic acid, acetylformic acid, alpha-ketopropionic acid, 2-ketopropionic acid, 2-oxopropanoate, propanoic acid, 2-oxo, 2-oxo-propionic acid, 2-oxopropansaeure | |
| CHEBI:32816 | |
| CC(=O)C(O)=O |
Safety and Handling
EINECSNumber : (2)-0959&(2)-1473
RTECSNumber : UZ0829800
TSCA : Yes
Recommended Storage : Refrigerator