Learn More
Pyruvic acid, sodium salt, 99+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 132150250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Pyruvic acid, sodium salt | |
>300.0°C | |
5.5 to 6.5 (10% soln. at 20°C) | |
Authentic | |
Plastic bottle | |
CH3COCO2Na | |
25 g | |
sodium pyruvate, pyruvic acid sodium salt, sodium 2-oxopropanoate, pyruvic acid, sodium salt, propanoic acid, 2-oxo-, sodium salt, sodium alpha-ketopropionate, pyruvate sodium, unii-pod38aif08, 2-oxopropanoic acid sodium salt, pod38aif08 | |
DAEPDZWVDSPTHF-UHFFFAOYSA-M | |
sodium;2-oxopropanoate | |
23662274 | |
110.04 | |
Solution |
113-24-6 | |
Colorless to Yellow | |
0.5% max. | |
99+% | |
C3H3NaO3 | |
MFCD00002586 | |
03, 608 | |
(10% in water) Clear colorless to light yellow | |
CC(=O)C(=O)[O-].[Na+] | |
110.04 | |
CHEBI:50144 | |
99+% |
Chemical Identifiers
113-24-6 | |
110.04 | |
DAEPDZWVDSPTHF-UHFFFAOYSA-M | |
23662274 | |
sodium;2-oxopropanoate |
C3H3NaO3 | |
MFCD00002586 | |
sodium pyruvate, pyruvic acid sodium salt, sodium 2-oxopropanoate, pyruvic acid, sodium salt, propanoic acid, 2-oxo-, sodium salt, sodium alpha-ketopropionate, pyruvate sodium, unii-pod38aif08, 2-oxopropanoic acid sodium salt, pod38aif08 | |
CHEBI:50144 | |
CC(=O)C(=O)[O-].[Na+] |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
EINECSNumber : 204-024-4
RUO – Research Use Only