missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pyrrolidine Hydrochloride, TCI America™

Manufacturer:  TCI America P24851G

 View more versions of this product

Catalog No. P24851G



Specifications

Specifications

Pyrrolidine Hydrochloride
1g
C4H10ClN
Pyrrolidinium Chloride
C1CCNC1.Cl
107.581
107.58
≥98.0% (T)
White
25150-61-2
MFCD01687885
FCLZCOCSZQNREK-UHFFFAOYSA-N
pyrrolidine;hydrochloride
212848
Crystal-Powder at 20°C

Chemical Identifiers

25150-61-2
107.581
FCLZCOCSZQNREK-UHFFFAOYSA-N
212848
C1CCNC1.Cl
C4H10ClN
MFCD01687885
Pyrrolidinium Chloride
pyrrolidine;hydrochloride
Safety and Handling

Safety and Handling

TSCA : No

Documents