missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pyrrolidine, 99%, Alfa Aesar™

Manufacturer:  Alfa Aesar A14852AP

 View more versions of this product

Catalog No. AAA14852AP



Description

Description

Applications
Pyrrolidine is used as building block in the synthesis of organic compounds. It is used to activate ketones and aldehydes toward nucleophilic addition. It is also utilized in promoting aldol condensation of ketones and aldehydes by forming enamine.

Solubility
Miscible with alcohol, ether, chloroform and water.

Notes
Store in cool place. Incompatible with acid chlorides, acid anhydrides, strong oxidizing agents, carbon dioxide and acids.
Specifications

Specifications

Pyrrolidine
500mL
99%
MFCD00005249
102395
tetrahydropyrrole, tetrahydro pyrrole, azacyclopentane, azolidine, tetramethylenimine, butylenimine, perhydropyrrole, prolamine, 1-azacyclopentane, tetramethyleneimine
RWRDLPDLKQPQOW-UHFFFAOYSA-N
pyrrolidine
31268
71.12
99%
0.856
123-75-1
C4H9N
UN1922
14,8015
Soluble in alcohol, ether, chloroform; Miscible with water
C1CCNC1
71.123
CHEBI:33135
Liquid

Chemical Identifiers

123-75-1
71.123
RWRDLPDLKQPQOW-UHFFFAOYSA-N
31268
pyrrolidine
C4H9N
MFCD00005249
tetrahydropyrrole, tetrahydro pyrrole, azacyclopentane, azolidine, tetramethylenimine, butylenimine, perhydropyrrole, prolamine, 1-azacyclopentane, tetramethyleneimine
CHEBI:33135
C1CCNC1
Safety and Handling

Safety and Handling

GHS H Statement
H225-H301-H314-H318-H332

GHS P Statement
P210-P260-P261-P280-P301+P310-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: PYRROLIDINE

RTECSNumber : UX9650000

TSCA : TSCA

storageNote1 : Air sensitive

SDS Documents