Learn More
Pyrrolidine, 99+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 132082500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Pyrrolidine | |
123-75-1 | |
0.8660g/mL | |
3°C | |
Glass bottle | |
1.4421 to 1.4441 | |
250 mL | |
01,972; 02,354; 03,240; 07,309; 08,428; 13,14 | |
15,8129 | |
Solubility in water: freely soluble. Other solubilities: soluble in alcohol, ether and chloroform, soluble in most organic solvents | |
[H+].[Cl-].C1CCNC1 | |
107.58 | |
0.94 mPa.s (20°C) | |
71.11 |
99+% | |
-63.0°C | |
86.0°C to 88.0°C | |
99% min. (GC) | |
C4H10ClN | |
MFCD00005249 | |
20,4 | |
0.866 | |
tetrahydropyrrole, tetrahydro pyrrole, azacyclopentane, azolidine, tetramethylenimine, butylenimine, perhydropyrrole, prolamine, 1-azacyclopentane, tetramethyleneimine | |
FCLZCOCSZQNREK-UHFFFAOYSA-N | |
pyrrolidine | |
31268 | |
CHEBI:33135 | |
99+% |
Chemical Identifiers
123-75-1 | |
107.58 | |
FCLZCOCSZQNREK-UHFFFAOYSA-N | |
31268 | |
[H+].[Cl-].C1CCNC1 |
C4H10ClN | |
MFCD00005249 | |
tetrahydropyrrole, tetrahydro pyrrole, azacyclopentane, azolidine, tetramethylenimine, butylenimine, perhydropyrrole, prolamine, 1-azacyclopentane, tetramethyleneimine | |
CHEBI:33135 |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.
Highly flammable liquid and vapor.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
EINECSNumber : 204-648-7
RTECSNumber : UX9650000
TSCA : TSCA
RUO – Research Use Only