missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Pyrrolidine, 99.5+%, purified by redistillation, AcroSeal™, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC446148000
Specifications
Pyrrolidine | |
3°C | |
1.4421 to 1.4441 | |
0.94 mPa.s (20°C) | |
-63.0°C | |
123-75-1 | |
C4H9N | |
1-azacyclopentane, azacyclopentane, azolidine, butylenimine, perhydropyrrole, prolamine, tetrahydro pyrrole, tetrahydropyrrole, tetramethyleneimine, tetramethylenimine | |
RWRDLPDLKQPQOW-UHFFFAOYSA-N | |
pyrrolidine | |
31268 | |
71.11 |
0.8660g/mL | |
AcroSeal™ Glass bottle | |
0.866 | |
86.0°C to 88.0°C | |
800mL | |
99.4% min. (GC) | |
15,8129 | |
Solubility in water: freely soluble. Other solubilities: soluble in alcohol, ether and chloroform, soluble in most organic solvents | |
C1CCNC1 | |
71.11 | |
CHEBI:33135 | |
99.5+% |
Chemical Identifiers
123-75-1 | |
71.11 | |
1-azacyclopentane, azacyclopentane, azolidine, butylenimine, perhydropyrrole, prolamine, tetrahydro pyrrole, tetrahydropyrrole, tetramethyleneimine, tetramethylenimine | |
CHEBI:33135 | |
C1CCNC1 |
C4H9N | |
RWRDLPDLKQPQOW-UHFFFAOYSA-N | |
31268 | |
pyrrolidine |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.
Highly flammable liquid and vapor.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
EINECSNumber : 204-648-7