Please login to your online account to display your discounted pricing

Alfa Aesar™ Pyrrolidine-2-thione, 97%

$115.57 - $479.15

Chemical Identifiers

Chemical Name or Material Pyrrolidine-2-thione
CAS 2295-35-4
Molecular Formula C4H7NS
Formula Weight 101.17
MDL Number MFCD00049351
Beilstein 105242
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0937403

  SDS
alfa aesar™
L0937403
1g Each for $115.57

AAL0937406

  SDS
alfa aesar™
L0937406
5g Each for $479.15
Description & Specifications

Specifications

Formula Weight 101.17
Chemical Name or Material Pyrrolidine-2-thione
Molecular Formula C4H7NS
Health Hazard 1 Warning
MDL Number MFCD00049351
CAS 2295-35-4
Beilstein 105242
Health Hazard 3 GHS P Statement
P280f
Wear protective clothing.
RTECS Number UX9647500
Quantity 1g
Assay 97%
Melting Point 109° to 113°C
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
Safety and Handling

Warning

GHS P Statement
P280f
Wear protective clothing.

GHS H Statement
H302
Harmful if swallowed.