Please login to your online account to display your discounted pricing

Pyrrolidine-1-carboximidamide hydroiodide, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  KM00683DA

 View more versions of this product

Catalog No. KM00683DA

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 241.07
Quantity 1g
Assay 97%
CAS Min % 97.0
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay Percent Range 97%
MDL Number MFCD00114285
Molecular Formula C5H11N3·HI
Packaging Amber Glass Bottle
Chemical Name or Material Pyrrolidine-1-carboximidamide hydroiodide
CAS 102392-83-6
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Pyrrolidine-1-carboximidamide hydroiodide
CAS 102392-83-6
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C5H11N3·HI
Formula Weight 241.07
MDL Number MFCD00114285
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents