missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pyrrolidine-1-carbonitrile, TCI America™

Manufacturer:  TCI America P23375G

 View more versions of this product

Catalog No. P23375G



Specifications

Specifications

Pyrrolidine-1-carbonitrile
Yellow
1530-88-7
MFCD00003165
1-pyrrolidinecarbonitrile, 1-cyanopyrrolidine, n-cyanopyrrolidine, pyrrolidinecarbonitrile, cyanopyrrolidine, cyano-pyrrolidine, tetramethylenecyanamide, 1-pyrrolidine carbonitrile, d00fga
C1CCN(C1)C#N
96.133
96.13
≥98.0% (GC)
110°C
5g
C5H8N2
3276
QJRYYOWARFCJQZ-UHFFFAOYSA-N
pyrrolidine-1-carbonitrile
73737
Clear Liquid at 20°C

Chemical Identifiers

1530-88-7
96.133
QJRYYOWARFCJQZ-UHFFFAOYSA-N
73737
C1CCN(C1)C#N
C5H8N2
MFCD00003165
1-pyrrolidinecarbonitrile, 1-cyanopyrrolidine, n-cyanopyrrolidine, pyrrolidinecarbonitrile, cyanopyrrolidine, cyano-pyrrolidine, tetramethylenecyanamide, 1-pyrrolidine carbonitrile, d00fga
pyrrolidine-1-carbonitrile
Safety and Handling

Safety and Handling

TSCA : Yes

Documents